Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentaerythrityl tetrabromide 98%

Supplier: AMBEED, INC
Warning

A248726-1G A248726-5G
77013-946EA 6 USD InStock
77013-946 77013-948
Pentaerythrityl tetrabromide 98%
Pentaerythrityl tetrabromide
Precaución: for Research Use Only


Formula: C₅H₈Br₄
MW: 387.73 g/mol
Boiling Pt: 305
Melting Pt: 163
MDL Number: MFCD00000210
CAS Number: 3229-00-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty