Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-1,2,4-triazole 97%

Supplier: AMBEED, INC
Warning

A249495-1G A249495-250MG A249495-5G A249495-10G
77000-628EA 10 USD InStock
77000-628 77000-630 77000-632 77672-224
3-Chloro-1,2,4-triazole 97%
3-Chloro-1,2,4-triazole
Precaución: for Research Use Only


Formula: C₂H₂ClN₃
MDL Number: MFCD00128812
CAS Number: 6818-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty