Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thieno[2,3-c]pyridine 95%

Supplier: AMBEED, INC
Warning

A250413-100MG A250413-1G A250413-250MG A250413-5G
77031-486EA 16 USD InStock
77031-486 77031-488 77031-490 77031-492
Thieno[2,3-c]pyridine 95%
Thieno[2,3-c]pyridine
Precaución: for Research Use Only


Formula: C₇H₅NS
MDL Number: MFCD01760492
CAS Number: 272-12-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty