Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,3S)-3-Aminocyclopentanol hydrochloride 95%

Supplier: AMBEED, INC
Warning

A250560-100MG A250560-1G A250560-250MG A250560-25G A250560-5G
76987-878EA 26 USD InStock
76987-878 76987-880 76987-882 76987-884 76987-886
(1S,3S)-3-Aminocyclopentanol hydrochloride 95%
(1S,3S)-3-Aminocyclopentanol hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClNO
MDL Number: MFCD23704943
CAS Number: 1523530-42-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty