Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-nitrobenzaldehyde 98%

Supplier: AMBEED, INC
Warning

A251367-10G A251367-1G A251367-250MG A251367-25G A251367-5G
77000-666EA 116 USD InStock
77000-666 77000-668 77000-670 77000-672 77000-674
2-Fluoro-4-nitrobenzaldehyde 98%
2-Fluoro-4-nitrobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₄FNO₃
MDL Number: MFCD00968941
CAS Number: 157701-72-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty