Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chlorooxazolo[4,5-b]pyridin-2(3H)-one 97%

Supplier: AMBEED, INC
Warning

A252393-10G A252393-1G A252393-5G
77031-560EA 35 USD InStock
77031-560 77031-562 77031-564 77031-566
6-Chlorooxazolo[4,5-b]pyridin-2(3H)-one 97%
6-Chlorooxazolo[4,5-b]pyridin-2(3H)-one
Precaución: for Research Use Only


Formula: C₆H₃ClN₂O₂
MDL Number: MFCD14706178
CAS Number: 35570-68-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty