Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-[3-(Trifluoromethyl)phenoxy]azetidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A252836-100MG A252836-1G
77031-580EA 30 USD InStock
77031-580 77031-582
3-[3-(Trifluoromethyl)phenoxy]azetidine hydrochloride 97%
3-[3-(Trifluoromethyl)phenoxy]azetidine hydrochloride
Precaución: for Research Use Only


Formula: C10H11ClF3NO
MDL Number: MFCD05664837
CAS Number: 1188374-88-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty