Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tanshinone IIA-sulfonic sodium 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A255311-10MG A255311-1MG A255311-250MG A255311-25MG A255311-50MG A255311-100MG
76988-006EA 12 USD InStock
76988-006 76988-008 76988-010 76988-012 76988-014 77683-770
Tanshinone IIA-sulfonic sodium 98%, Ambeed.Inc
Tanshinone IIA-sulfonic sodium
Precaución: For research use only.


Formula: C₁₉H₁₇NaO₆S
MDL Number: MFCD09028094
CAS Number: 69659-80-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty