Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 3,4-dihydroxythiophene-2,5-dicarboxylate 95%

Supplier: AMBEED, INC
Warning

A255964-1G A255964-250MG A255964-25G A255964-5G
77014-078EA 20 USD InStock
77014-078 77014-080 77014-082 77014-084
Diethyl 3,4-dihydroxythiophene-2,5-dicarboxylate 95%
Diethyl 3,4-dihydroxythiophene-2,5-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₆S
MW: 260.264 g/mol
MDL Number: MFCD03094035
CAS Number: 1822-66-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty