Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine 97%

Supplier: AMBEED, INC
Warning

A256245-250MG A256245-25G A256245-1G A256245-10G
77000-828EA 15 USD InStock
77000-828 77000-830 77672-288 77672-286
3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine 97%
3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine
Precaución: for Research Use Only


Formula: C₁₁H₁₆ClN₃
MDL Number: MFCD01951505
CAS Number: 16154-72-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty