Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-mercaptobenzothiazole 98%

Supplier: AMBEED, INC
Warning

A256442-100MG A256442-1G A256442-250MG A256442-5G
77000-832EA 6 USD InStock
77000-832 77000-834 77000-836 77000-838
5-Fluoro-2-mercaptobenzothiazole 98%
5-Fluoro-2-mercaptobenzothiazole
Precaución: for Research Use Only


Formula: C₇H₄FNS₂
MDL Number: MFCD02181177
CAS Number: 155559-81-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty