Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2,6-dichloro-3-methylisonicotinate 97%

Supplier: AMBEED, INC
Warning

A256607-100MG A256607-1G A256607-250MG A256607-5G
76988-060EA 15 USD InStock
76988-060 76988-062 76988-064 76988-066
Ethyl 2,6-dichloro-3-methylisonicotinate 97%
Ethyl 2,6-dichloro-3-methylisonicotinate
Precaución: for Research Use Only


Formula: C₉H₉Cl₂NO₂
MDL Number: MFCD05156546
CAS Number: 137520-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty