Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dichloro-D-phenylalanine 97%

Supplier: AMBEED, INC
Warning

A256968-10G A256968-1G A256968-250MG
77000-850EA 223 USD InStock
77000-850 77000-852 77000-854
2,4-Dichloro-D-phenylalanine 97%
2,4-Dichloro-D-phenylalanine
Precaución: for Research Use Only


Formula: C₉H₉Cl₂NO₂
MDL Number: MFCD01860873
CAS Number: 114872-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty