Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A258582-100MG A258582-1G A258582-250MG A258582-5G
76988-132EA 39 USD InStock
76988-132 76988-134 76988-136 76988-138
3-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid 98%
3-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₅BFNO₃
MDL Number: MFCD08235060
CAS Number: 874219-43-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty