Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Fluoropyridin-2-yl)methanamine dihydrochloride 95% (contains 3-7% NH4Cl)

Supplier: AMBEED, INC
Warning

A261138-1G A261138-250MG
76988-190EA 16 USD InStock
76988-190 76988-192
(3-Fluoropyridin-2-yl)methanamine dihydrochloride 95% (contains 3-7% NH4Cl)
(3-Fluoropyridin-2-yl)methanamine dihydrochloride
Precaución: for Research Use Only


Formula: C₆H₉Cl₂FN₂
MDL Number: MFCD09954760
CAS Number: 312904-49-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty