Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[3,2-c]pyridine-7-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A261524-100MG A261524-1G A261524-50MG
76988-194EA 51 USD InStock
76988-194 76988-196 76988-198
1H-Pyrrolo[3,2-c]pyridine-7-carboxylic acid 97%
1H-Pyrrolo[3,2-c]pyridine-7-carboxylic acid
Precaución: for Research Use Only


Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
MDL Number: MFCD11845522
CAS Number: 1082040-92-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty