Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-((2,4-Dioxothiazolidin-5-ylidene)methyl)-2-methoxyphenoxy)-3-(trifluoromethyl)benzonitrile 95%

Supplier: AMBEED, INC
Warning

A262648-100MG A262648-250MG
77014-214EA 52 USD InStock
77014-214 77014-216
4-(4-((2,4-Dioxothiazolidin-5-ylidene)methyl)-2-methoxyphenoxy)-3-(trifluoromethyl)benzonitrile 95%
4-(4-((2,4-Dioxothiazolidin-5-ylidene)methyl)-2-methoxyphenoxy)-3-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₁₉H₁₁F₃N₂O₄S
MW: 420.36 g/mol
MDL Number: MFCD29034874
CAS Number: 1055361-35-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty