Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromaminic acid 98%

Supplier: AMBEED, INC
Warning

A265473-100G A265473-25G A265473-5G
77001-018EA 52 USD InStock
77001-018 77001-020 77001-024
Bromaminic acid 98%
Bromaminic acid
Precaución: for Research Use Only


Formula: C₁₄H₈BrNO₅S
MDL Number: MFCD00035694
CAS Number: 116-81-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty