Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Di(thiophen-2-yl)dithieno[3,2-b:2',3'-d]thiophene 95+%

Supplier: AMBEED, INC
Warning

A265542-100MG A265542-25MG A265542-50MG
77014-276EA 192 USD InStock
77014-276 77014-278 77014-280
2,6-Di(thiophen-2-yl)dithieno[3,2-b:2',3'-d]thiophene 95+%
2,6-Di(thiophen-2-yl)dithieno[3,2-b:2',3'-d]thiophene
Precaución: for Research Use Only


Formula: C₁₆H₈S₅
MDL Number: MFCD11215198
CAS Number: 910788-24-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty