Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3-hydroxyphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A265828-1G A265828-250MG A265828-5G
77031-844EA 18 USD InStock
77031-844 77031-846 77031-848
2,6-Difluoro-3-hydroxyphenylboronic acid 97%
2,6-Difluoro-3-hydroxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BF₂O₃
MW: 173.91 g/mol
MDL Number: MFCD09878345
CAS Number: 957065-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty