Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methylazepan-4-one hydrochloride 98%

Supplier: AMBEED, INC
Warning

A268552-1G A268552-25G A268552-5G
77001-062EA 15 USD InStock
77001-062 77001-064 77001-066
1-Methylazepan-4-one hydrochloride 98%
1-Methylazepan-4-one hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₄ClNO
MDL Number: MFCD07779465
CAS Number: 19869-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty