Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-methyl-6-phenylpicolinate 95%

Supplier: AMBEED, INC
Warning

A268846-100MG A268846-1G A268846-250MG
77014-370EA 9 USD InStock
77014-370 77014-372 77014-374
Methyl 4-methyl-6-phenylpicolinate 95%
Methyl 4-methyl-6-phenylpicolinate
Precaución: for Research Use Only


Formula: C₁₄H₁₃NO₂
MW: 227.26 g/mol
MDL Number: MFCD27938857
CAS Number: 119715-66-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty