Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-nitro-1H-pyrazole 96%

Supplier: AMBEED, INC
Warning

A269271-100MG A269271-1G A269271-250MG A269271-5G
76988-294EA 39 USD InStock
76988-294 76988-296 76988-298 76988-300
3-Bromo-4-nitro-1H-pyrazole 96%
3-Bromo-4-nitro-1H-pyrazole
Precaución: for Research Use Only


Formula: C₃H₂BrN₃O₂
MDL Number: MFCD19441764
CAS Number: 784193-37-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty