Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Enzalutamide 98%

Supplier: AMBEED, INC
Warning

A270395-100MG A270395-10MG A270395-1G A270395-1MG A270395-250MG A270395-25G A270395-25MG A270395-50MG A270395-5G A270395-5MG
77031-904EA 16 USD InStock
77031-904 77031-906 77031-908 77031-910 77031-912 77031-914 77031-916 77031-918 77031-920 77031-922
Enzalutamide 98%
Enzalutamide
Precaución: for Research Use Only


Formula: C₂₁H₁₆F₄N₄O₂S
MW: 464.44 g/mol
MDL Number: MFCD14155804
CAS Number: 915087-33-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty