Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-4-phenyl-1H-indene 97%

Supplier: AMBEED, INC
Warning

A273149-100MG A273149-1G A273149-250MG A273149-5G
77014-454EA 7 USD InStock
77014-454 77014-456 77014-458 77014-460
2-Methyl-4-phenyl-1H-indene 97%
2-Methyl-4-phenyl-1H-indene
Precaución: for Research Use Only


Formula: C₁₆H₁₄
MDL Number: MFCD06797435
CAS Number: 159531-97-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty