Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl gold(I) chloride 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A277202-100MG A277202-250MG A277202-1G
76988-426EA 28 USD InStock
76988-426 76988-428 77683-837
2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl gold(I) chloride 98%, Ambeed.Inc
2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl gold(I) chloride
Precaución: For research use only.


Formula: C₃₃H₄₉AuClP
MW: 709.14 g/mol
MDL Number: MFCD09842765
CAS Number: 854045-94-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty