Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-(2-fluoro-4-nitrophenyl)piperazine 95%

Supplier: AMBEED, INC
Warning

A278790-10G A278790-1G A278790-250MG
77002-106EA 211 USD InStock
77002-106 77002-108 77002-110
1-Boc-4-(2-fluoro-4-nitrophenyl)piperazine 95%
1-Boc-4-(2-fluoro-4-nitrophenyl)piperazine
Precaución: for Research Use Only


Formula: C₁₅H₂₀FN₃O₄
MW: 325.34 g/mol
MDL Number: MFCD08443959
CAS Number: 154590-34-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty