Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-glutamine 98%

Supplier: AMBEED, INC
Warning

A286579-100G A286579-1G A286579-25G A286579-5G
77002-268EA 223 USD InStock
77002-268 77002-270 77002-272 77002-274
Fmoc-D-glutamine 98%
Fmoc-D-glutamine
Precaución: for Research Use Only


Formula: C₂₀H₂₀N₂O₅
MW: 368.389 g/mol
MDL Number: MFCD00062959
CAS Number: 112898-00-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty