Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dichloro-4-iodopyridine 95%

Supplier: AMBEED, INC
Danger

A287606-100MG A287606-10G A287606-1G A287606-250MG A287606-25G A287606-5G
77002-282EA 5 USD InStock
77002-282 77002-284 77002-286 77002-288 77002-290 77002-292
2,3-Dichloro-4-iodopyridine 95%
2,3-Dichloro-4-iodopyridine
Precaución: for Research Use Only


Formula: C₅H₂Cl₂IN
MW: 273.8900 g/mol
MDL Number: MFCD11857737
CAS Number: 889865-45-6
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty