Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-(trifluoromethyl)benzonitrile 96%

Supplier: AMBEED, INC
Warning

A287906-1G A287906-250MG
77032-242EA 45 USD InStock
77032-242 77032-244
2-Bromo-3-(trifluoromethyl)benzonitrile 96%
2-Bromo-3-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₃BrF₃N
MW: 250.02 g/mol
MDL Number: MFCD08532491
CAS Number: 914637-07-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty