Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2-Nitrophenyl)methanamine 98%

Supplier: AMBEED, INC
Danger

A290163-100MG A290163-250MG
77032-276EA 53 USD InStock
77032-276 77032-278
(2-Nitrophenyl)methanamine 98%
(2-Nitrophenyl)methanamine
Precaución: for Research Use Only


Formula: C₇H₈N₂O₂
MDL Number: MFCD03411002
CAS Number: 1904-78-5
UN: 2735
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty