Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzylamino)-2-((3-chloro-4-hydroxyphenethyl)amino)pyrimidine-5-carboxamide ≥95%

Supplier: AMBEED, INC
Warning

A290303-10MG A290303-1MG A290303-5MG
76988-644EA 124 USD InStock
76988-644 76988-646 76988-648
4-(Benzylamino)-2-((3-chloro-4-hydroxyphenethyl)amino)pyrimidine-5-carboxamide ≥95%
4-(Benzylamino)-2-((3-chloro-4-hydroxyphenethyl)amino)pyrimidine-5-carboxamide
Precaución: for Research Use Only


Formula: C₂₀H₂₀ClN₅O₂
MW: 397.86 g/mol
MDL Number: MFCD24038757
CAS Number: 919486-40-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty