Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromonicotinic acid 98%

Supplier: AMBEED, INC
Warning

A291101-250MG A291101-1G
77002-356EA 15 USD InStock
77002-356 77672-490
4-Bromonicotinic acid 98%
4-Bromonicotinic acid
Precaución: for Research Use Only


Formula: C₆H₄BrNO₂
MW: 202.007 g/mol
MDL Number: MFCD04114236
CAS Number: 15366-62-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty