Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Z-Val-OEt 95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A291268-10G A291268-1G A291268-5G A291268-25G
77014-786EA 41 USD InStock
77014-786 77014-788 77014-790 77683-874
Z-Val-OEt 95%, Ambeed.Inc
Z-Val-OEt
Precaución: For research use only.


Formula: C₁₅H₂₁NO₄
MDL Number: MFCD06658546
CAS Number: 67436-18-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty