Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2-Amino-3-benzyloxy-1-propanol 97%

Supplier: AMBEED, INC
Danger

A292604-10G A292604-1G
77032-330EA 285 USD InStock
77032-330 77571-806
(R)-(+)-2-Amino-3-benzyloxy-1-propanol 97%
(R)-(+)-2-Amino-3-benzyloxy-1-propanol
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₂
MW: 181.235 g/mol
MDL Number: MFCD00235943
CAS Number: 58577-87-0
UN: 3267
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty