Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-1,2-Cyclohexanedimethanol 98%

Supplier: AMBEED, INC
Warning

A292674-100MG A292674-1G A292674-250MG
76988-674EA 18 USD InStock
76988-674 76988-676 76988-678
trans-1,2-Cyclohexanedimethanol 98%
trans-1,2-Cyclohexanedimethanol
Precaución: for Research Use Only


Formula: C₈H₁₆O₂
MDL Number: MFCD09260493
CAS Number: 25712-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty