Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-fluoro-3-nitrobenzotrifluoride 97%

Supplier: AMBEED, INC
Warning

A294552-100MG A294552-1G A294552-250MG A294552-5G
77032-374EA 8 USD InStock
77032-374 77032-376 77032-378 77032-380
2-Amino-5-fluoro-3-nitrobenzotrifluoride 97%
2-Amino-5-fluoro-3-nitrobenzotrifluoride
Precaución: for Research Use Only


Formula: C₇H₄F₄N₂O₂
MDL Number: MFCD03094266
CAS Number: 344-29-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty