Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Phenyl-1-hydroxy-2-naphthoate 98%

Supplier: AMBEED, INC
Warning

A298464-25G A298464-5G
77002-468EA 44 USD InStock
77002-468 77002-470
Phenyl-1-hydroxy-2-naphthoate 98%
Phenyl-1-hydroxy-2-naphthoate
Precaución: for Research Use Only


Formula: C₁₇H₁₂O₃
MW: 264.28 g/mol
Boiling Pt: 414
Melting Pt: 96
MDL Number: MFCD00014310
CAS Number: 132-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty