Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-6-bromopyrazine-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A298697-100MG A298697-10G A298697-1G A298697-250MG A298697-5G
77032-436EA 9 USD InStock
77032-436 77032-438 77032-440 77032-442 77032-444
Methyl-6-bromopyrazine-2-carboxylate 98%
Methyl-6-bromopyrazine-2-carboxylate
Precaución: for Research Use Only


Formula: C₆H₅BrN₂O₂
MW: 217.0200 g/mol
MDL Number: MFCD14155890
CAS Number: 40155-34-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty