Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dibromo-3-fluoropyridine 98%

Supplier: AMBEED, INC
Warning

A298873-100MG A298873-10G A298873-1G A298873-250MG A298873-25G
77002-472EA 5 USD InStock
77002-472 77002-474 77002-476 77002-478 77002-480
2,5-Dibromo-3-fluoropyridine 98%
2,5-Dibromo-3-fluoropyridine
Precaución: for Research Use Only


Formula: C₅H₂Br₂FN
MDL Number: MFCD06659497
CAS Number: 156772-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty