Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-(dodecyloxy)-2-oxoethanesulfonate 97%

Supplier: AMBEED, INC
Warning

A299187-100G A299187-25G A299187-5G
77032-454EA 179 USD InStock
77032-454 77032-456 77032-458
Sodium 2-(dodecyloxy)-2-oxoethanesulfonate 97%
Sodium 2-(dodecyloxy)-2-oxoethanesulfonate
Precaución: for Research Use Only


Formula: C₁₄H₂₇NaO₅S
MDL Number: MFCD00214054
CAS Number: 1847-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty