Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A299206-1G A299206-250MG A299206-5G
76988-740EA 32 USD InStock
76988-740 76988-742 77683-893
1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene 98%, Ambeed.Inc
1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene
Precaución: For research use only.


Formula: C₂₇H₃₆N₂
MDL Number: MFCD07781991
CAS Number: 244187-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty