Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-(3-(Diisopropylamino)-1-phenylpropyl)-4-methylphenol (2R,3R)-2,3-dihydroxysuccinate 98%

Supplier: AMBEED, INC
Warning

A299510-100MG A299510-250MG A299510-25G A299510-50MG A299510-5G A299510-10MG
77002-492EA 8 USD InStock
77002-492 77002-494 77002-496 77002-498 77002-500 77672-526
(R)-2-(3-(Diisopropylamino)-1-phenylpropyl)-4-methylphenol (2R,3R)-2,3-dihydroxysuccinate 98%
(R)-2-(3-(Diisopropylamino)-1-phenylpropyl)-4-methylphenol (2R,3R)-2,3-dihydroxysuccinate
Precaución: for Research Use Only


Formula: C₂₆H₃₇NO₇
MDL Number: MFCD07771985
CAS Number: 124937-52-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty