Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Combretastatin A4 98%

Supplier: AMBEED, INC
Danger

A300403-100MG A300403-10MG A300403-1G A300403-1MG A300403-250MG A300403-25MG A300403-5MG A300403-5G
77002-504EA 94 USD InStock
77002-504 77002-506 77002-508 77002-510 77002-512 77002-514 77002-516 77672-528
Combretastatin A4 98%
Combretastatin A4
Precaución: for Research Use Only


Formula: C₁₈H₂₀O₅
MDL Number: MFCD03453309
CAS Number: 117048-59-6
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty