Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2,3-diphenylpyrazine 98%

Supplier: AMBEED, INC
Warning

A304104-100G A304104-1G A304104-250MG A304104-25G A304104-5G
76988-834EA 86 USD InStock
76988-834 76988-836 76988-838 76988-840 76988-842
5-Chloro-2,3-diphenylpyrazine 98%
5-Chloro-2,3-diphenylpyrazine
Precaución: for Research Use Only


Formula: C₁₆H₁₁ClN₂
MDL Number: MFCD00234892
CAS Number: 41270-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty