Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxyquinoline-3-carbaldehyde 98+%

Supplier: AMBEED, INC
Danger

A307387-1G A307387-250MG A307387-5G
77002-608EA 42 USD InStock
77002-608 77002-610 77002-612
2-Methoxyquinoline-3-carbaldehyde 98+%
2-Methoxyquinoline-3-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₉NO₂
MW: 187.2 g/mol
MDL Number: MFCD03864951
CAS Number: 139549-06-7
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty