Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl 1H-indole-2,5-dicarboxylate 95%

Supplier: AMBEED, INC
Warning

A307922-100MG A307922-1G A307922-250MG
77002-640EA 15 USD InStock
77002-640 77002-642 77002-644
Diethyl 1H-indole-2,5-dicarboxylate 95%
Diethyl 1H-indole-2,5-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₅NO₄
MW: 261.27 g/mol
MDL Number: MFCD03094964
CAS Number: 127221-02-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty