Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl thiazole-5-carboxylate 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A308629-10G A308629-1G A308629-5G A308629-25G
77015-106EA 75 USD InStock
77015-106 77015-108 77015-110 77683-910
Methyl thiazole-5-carboxylate 98%, Ambeed.Inc
Methyl thiazole-5-carboxylate
Precaución: For research use only.


Formula: C₅H₅NO₂S
MDL Number: MFCD03788563
CAS Number: 14527-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty