Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gallic acid 98%

Supplier: AMBEED, INC
Warning

A311911-1000G A311911-100G A311911-25G A311911-500G A311911-50MG
77002-698EA 49 USD InStock
77002-698 77002-700 77002-702 77002-704 77002-706 77672-594
Gallic acid 98%
Gallic acid
Precaución: for Research Use Only


Formula: C₇H₆O₅
MW: 170.12 g/mol
Boiling Pt: 501
MDL Number: MFCD00002510
CAS Number: 149-91-7
Merck Index: 13,04366
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty