Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-3-piperidineacetic acid 95%

Supplier: AMBEED, INC
Warning

A312891-10G A312891-1G A312891-250MG A312891-5G
77002-712EA 65 USD InStock
77002-712 77002-714 77002-716 77002-718
1-Boc-3-piperidineacetic acid 95%
1-Boc-3-piperidineacetic acid
Precaución: for Research Use Only


Formula: C₁₂H₂₁NO₄
MW: 243.3000 g/mol
MDL Number: MFCD02178999
CAS Number: 183483-09-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty